
Castalagin

Synonym(s):
(33β)-Vescalagin, NSC 297535
Empirical Formula (Hill Notation):
C41H26O26
CAS Number:
Molecular Weight:
934.63
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing