
Carvedilol impurity C

Synonym(s):
(2RS)-1-{Benzyl[2-(2-methoxyphenoxy)ethyl]amino}-3-(9H-carbazol-4-yloxy)-2-propanol, N-Benzylcarvedilol
Empirical Formula (Hill Notation):
C31H32N2O4
CAS Number:
Molecular Weight:
496.60
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing