
CAPSO sodium salt

Synonym(s):
3-(Cyclohexylamino)-2-hydroxy-1-propanesulfonic acid sodium salt
Linear Formula:
C9H18NO4SNa
CAS Number:
Molecular Weight:
259.30
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
pK Value
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing