
Calcium sulfate dihydrate

Synonym(s):
Calcium sulfate dihydrate
Linear Formula:
CaSO4 · 2H2O
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing