
Calcium Ionophore A23187 hemimagnesium salt

Synonym(s):
A23187 hemimagnesium salt, Antibiotic A 23187 hemimagnesium salt, Calcimycin hemimagnesium salt, Calimycin hemimagnesium salt
Empirical Formula (Hill Notation):
C29H36Mg0.5N3O6
CAS Number:
Molecular Weight:
534.77
Products
Brand
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing