
Budesonide impurity standard

Synonym(s):
Budesonide, 16,17-Butylidenebis(oxy)-11,21-dihydroxypregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C25H34O6
CAS Number:
Molecular Weight:
430.53
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing