
Bromazepam

Synonym(s):
7-Bromo-5-(2-pyridyl)-3H-1,4-benzodiaxepin-2(1H)-one
Empirical Formula (Hill Notation):
C14H10BrN3O
CAS Number:
Molecular Weight:
316.15
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing