
(Boc-Cys-OH)2

Synonym(s):
Nα,Nα′-di-Boc-L-cystine
Empirical Formula (Hill Notation):
C16H28N2O8S2
Products
Brand
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing