
bisBenzimide H 33342 trihydrochloride

Synonym(s):
2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342, Hoechst 33342, bisBenzimide
Empirical Formula (Hill Notation):
C27H28N6O · 3HCl · xH2O
CAS Number:
Molecular Weight:
561.93 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing