
Bis(4-fluorophenyl) sulfone

Synonym(s):
4,4′-Difluorodiphenyl sulfone, 4-Fluorophenyl sulfone, Bis(4-fluorophenyl) sulfone
Linear Formula:
(FC6H4)2SO2
CAS Number:
Molecular Weight:
254.25
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing