
Bis(4-chlorophenyl) sulfone

Synonym(s):
4,4′-Dichlorodiphenyl sulfone, 4-Chlorophenyl sulfone
Linear Formula:
(ClC6H4)2SO2
CAS Number:
Molecular Weight:
287.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2