
Benzocaine

Synonym(s):
4-Aminobenzoic acid ethyl ester, Ethyl 4-aminobenzoate
Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Products
Brand
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4