
Benazepril Related Compound C

Synonym(s):
(3S)-3-[[(1S)-1-Carboxy-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid, Benazeprilat
Empirical Formula (Hill Notation):
C22H24N2O5
CAS Number:
Molecular Weight:
396.44
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2