Skip to Content
Merck
Atto 590 NHS ester BioReagent, suitable for fluorescence, ≥90.0% (degree of coupling)

Atto 590 NHS ester

Atto 590 NHS ester BioReagent, suitable for fluorescence, ≥90.0% (degree of coupling)
Empirical Formula (Hill Notation):
C41H42ClN3O11
CAS Number:
Molecular Weight:
788.24

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Detection Method
Showing 1-1 of 1