Skip to Content
Merck
Ascentis® Express Biphenyl Guard Column pkg of 3 ea, 2.7 μm particle size, L × I.D. 0.5 cm × 4.6 mm

Ascentis® Express 90 Å Biphenyl (2.7 µm) HPLC Columns

Ascentis® Express Biphenyl Guard Column pkg of 3 ea, 2.7 μm particle size, L × I.D. 0.5 cm × 4.6 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-19 of 19
Compare
Product Number
Description
Pricing