
Arginine ethyl ester dihydrochloride

Synonym(s):
L-Arginine ethyl ester dihydrochloride
Empirical Formula (Hill Notation):
C8H18N4O2 · 2HCl
CAS Number:
Molecular Weight:
275.18
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing