
Anti-BRD1 antibody produced in rabbit

Synonym(s):
Anti-BR140-like protein antibody produced in rabbit, Anti-Bromodomain-containing protein 1 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing