
Antibodix® WCX-NP (1.7 μm) Weak Cation Exchange HPLC Columns

Products
Product Category
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing