Acetoacetyl coenzyme A sodium salt hydrate
Synonym(s):
Acetoacetyl-CoANa3
Empirical Formula (Hill Notation):
C25H40N7O18P3S · xNa+ · yH2O
CAS Number:
Molecular Weight:
851.61 (anhydrous free acid basis)
Products
Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing