
Acamprosate Related Compound A

Synonym(s):
3-Amino-1-propanesulfonic acid, 3APS, Homotaurine, Tramiprosate
Linear Formula:
H2N(CH2)3SO3H
CAS Number:
Molecular Weight:
139.17
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2