Skip to Content
Merck
7-Fluoro-1H-indole 97%

7-Fluoro-1H-indole

7-Fluoro-1H-indole 97%
Empirical Formula (Hill Notation):
C8H6FN
CAS Number:
Molecular Weight:
135.14

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing