
6,6′-Dinicotinic acid

Synonym(s):
2,2′-Bipyridine-5,5′-dicarboxylic acid, 6,6′-Binicotinic acid
Empirical Formula (Hill Notation):
C12H8N2O4
CAS Number:
Molecular Weight:
244.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing