Skip to Content
Merck
4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride technical grade

4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride

4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride technical grade
Empirical Formula (Hill Notation):
C11H13N · HCl
CAS Number:
Molecular Weight:
195.69
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing