Skip to Content
Merck
4-Methoxyphenylboronic acid ≥95.0%

4-Methoxyphenylboronic acid

4-Methoxyphenylboronic acid ≥95.0%
Synonym(s):
(4-Methoxyphenyl)boric acid, (p-Methoxyphenyl)boronic acid, 4-Anisylboronic acid, 4-Methoxybenzeneboronic acid, p-Anisylboronic acid, p-Methoxybenzeneboronic acid
Linear Formula:
CH3OC6H4B(OH)2
CAS Number:
Molecular Weight:
151.96

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing