Skip to Content
Merck
4-Bromo-2,6-dimethylaniline 98%

4-Bromo-2,6-dimethylaniline

4-Bromo-2,6-dimethylaniline 98%
Synonym(s):
4-Bromo-2,6-xylidine
Linear Formula:
BrC6H2(CH3)2NH2
CAS Number:
Molecular Weight:
200.08
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing