Skip to Content
Merck
4′-Aminobenzo-15-crown-5 97%

4′-Aminobenzo-15-crown-5

4′-Aminobenzo-15-crown-5 97%
Synonym(s):
(Benzo-15-crown-5)-4′-ylamine
Empirical Formula (Hill Notation):
C14H21NO5
CAS Number:
Molecular Weight:
283.32

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing