3-Methyl-1,5-pentanediol
Linear Formula:
HOCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
118.17
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing