Skip to Content
Merck
3-Bromo-5-methoxypyridine 97%

3-Bromo-5-methoxypyridine

3-Bromo-5-methoxypyridine 97%
Empirical Formula (Hill Notation):
C6H6BrNO
CAS Number:
Molecular Weight:
188.02

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing