Skip to Content
Merck
2-(Trimethylsilyl)ethoxymethyl chloride ≥95.0% (GC)

2-(Trimethylsilyl)ethoxymethyl chloride

2-(Trimethylsilyl)ethoxymethyl chloride ≥95.0% (GC)
Synonym(s):
(2-Chloromethoxyethyl)trimethylsilane, Chloromethyl 2-trimethylsilylethyl ether, SEM-Cl, SEM-chloride
Linear Formula:
(CH3)3SiCH2CH2OCH2Cl
CAS Number:
Molecular Weight:
166.72
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2