Skip to Content
Merck
(2S-cis)-(−)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid analytical standard

(2S-cis)-(−)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid

(2S-cis)-(−)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid analytical standard
Empirical Formula (Hill Notation):
C13H14N2O4
CAS Number:
Molecular Weight:
262.26
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique