
2-Methylcyclopropanecarboxylic acid

Linear Formula:
CH3C3H4CO2H
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing