
2-Iodo-1-methyl-1H-imidazole

Empirical Formula (Hill Notation):
C4H5IN2
CAS Number:
Molecular Weight:
208.00
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing