
2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers

Synonym(s):
2-Ethyl-3(5 or 6)-dimethylpyrazine, mixture of isomers
Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing