Skip to Content
Merck
2-Ethyl-2-phenylmalonamide monohydrate 99%

2-Ethyl-2-phenylmalonamide monohydrate

2-Ethyl-2-phenylmalonamide monohydrate 99%
Synonym(s):
PEMA
Linear Formula:
C6H5C(C2H5)(CONH2)2·H2O
CAS Number:
Molecular Weight:
224.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing