
2′-Deoxycytidine hydrochloride

Synonym(s):
1-(2-Deoxy-β-D-ribofuranosyl)cytosine hydrochloride, Cytosine deoxyriboside hydrochloride
Empirical Formula (Hill Notation):
C9H13N3O4 · HCl
CAS Number:
Molecular Weight:
263.68
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing