Skip to Content
Merck
2-Bromo-2-methylpropane 98%

2-Bromo-2-methylpropane

2-Bromo-2-methylpropane 98%
Synonym(s):
2-methyl-2-bromopropane, tert-Butyl bromide, 2-Bromo-2-methylpropane
Linear Formula:
(CH3)3CBr
CAS Number:
Molecular Weight:
137.02

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2