2-Benzylaniline
Synonym(s):
2-Aminodiphenylmethane
Linear Formula:
C6H5CH2C6H4NH2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing