
2-(5-Nitro-1H-imidazol-1-yl)ethanol

Synonym(s):
5-Nitro-1H-imidazole-1-ethanol
Empirical Formula (Hill Notation):
C5H7N3O3
CAS Number:
Molecular Weight:
157.13
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing