Skip to Content
Merck
2,4-Dichloroaniline PESTANAL®, analytical standard

2,4-Dichloroaniline

2,4-Dichloroaniline PESTANAL®, analytical standard
Linear Formula:
Cl2C6H3NH2
CAS Number:
Molecular Weight:
162.02
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing