![2,2,2-Trichloro-1,1-dimethylethyl chloroformate 96%](/deepweb/assets/sigmaaldrich/product/structures/222/344/6f7c906a-1544-40a4-995d-6986b06e3476/640/6f7c906a-1544-40a4-995d-6986b06e3476.png)
2,2,2-Trichloro-1,1-dimethylethyl chloroformate
![2,2,2-Trichloro-1,1-dimethylethyl chloroformate 96%](/deepweb/assets/sigmaaldrich/product/structures/222/344/6f7c906a-1544-40a4-995d-6986b06e3476/640/6f7c906a-1544-40a4-995d-6986b06e3476.png)
Synonym(s):
β,β,β-Trichloro-tert-butyl chloroformate, β,β,β-Trichloro-tert-butoxycarbonyl chloride, TCBoc-chloride
Linear Formula:
ClCO2C(CH3)2CCl3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing