Skip to Content
Merck
1-Oleoyl-2-acetyl-sn-glycerol ≥97% (TLC), oil

1-Oleoyl-2-acetyl-sn-glycerol

1-Oleoyl-2-acetyl-sn-glycerol ≥97% (TLC), oil
Synonym(s):
1-Oleoyl-2-acetyl-sn-glycerol, (9Z)-9-Octadecenoic acid (2S)-2-(acetyloxy)-3-hydroxypropyl ester, 1-(cis-9-Octadecenoyl)-2-acetyl-sn-glycerol, 2-Acetyl-1-oleoyl-sn-glycerol, DG(18:1(9Z)/2:0/0:0), OAG
Empirical Formula (Hill Notation):
C23H42O5
CAS Number:
Molecular Weight:
398.58

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Format
Research Area
Quality Segment
Functional Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing