Skip to Content
Merck
1H,1H-Perfluorooctyl methacrylate contains MEHQ as inhibitor, 96%

1H,1H-Perfluorooctyl methacrylate

1H,1H-Perfluorooctyl methacrylate contains MEHQ as inhibitor, 96%
Synonym(s):
1,1-Dihydroperfluorooctyl methacrylate, 1H,1H-Pentadecafluorooctyl methacrylate, 1H,H-Pentadecafluoro-1-octyl methacrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octyl methacrylate, Methacrylic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, Nyebar
Empirical Formula (Hill Notation):
C12H7F15O2
CAS Number:
Molecular Weight:
468.16
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing