Skip to Content
Merck
1-Butyl-3-methylimidazolium bromide >97.0% (HPLC)

1-Butyl-3-methylimidazolium bromide

1-Butyl-3-methylimidazolium bromide >97.0% (HPLC)
Synonym(s):
BMIMBr
Empirical Formula (Hill Notation):
C8H15BrN2
CAS Number:
Molecular Weight:
219.12

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2