Skip to Content
Merck
1-Aza-12-crown-4 ≥97.0%

1-Aza-12-crown-4

1-Aza-12-crown-4 ≥97.0%
Synonym(s):
1,4,7-Trioxa-10-azacyclododecane
Empirical Formula (Hill Notation):
C8H17NO3
CAS Number:
Molecular Weight:
175.23
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing