
1,6-Anhydro-D-glucose

Synonym(s):
1,6-Anhydro-β-D-glucose, Levoglucosan
Empirical Formula (Hill Notation):
C6H10O5
CAS Number:
Molecular Weight:
162.14
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing