Skip to Content
Merck
1,3-Bis(benzyloxycarbonyl)-2-methyl-2-thiopseudourea 95%

1,3-Bis(benzyloxycarbonyl)-2-methyl-2-thiopseudourea

1,3-Bis(benzyloxycarbonyl)-2-methyl-2-thiopseudourea 95%
Synonym(s):
1,3-Bis(benzyloxycarbonyl)-2-methylisothiourea, 2-Methyl-1,3-di-Z-isothiourea
Linear Formula:
C6H5CH2O2CN=C(SCH3)NHCO2CH2C6H5
CAS Number:
Molecular Weight:
358.41

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing