
10-Formylfolic acid

Synonym(s):
Formylfolic acid, N-[4-[[(2-Amino-1,4-dihydro-4-oxo-6-pteridinyl)methyl]formylamino]benzoyl]-L-glutamic acid
Empirical Formula (Hill Notation):
C20H19N7O7
CAS Number:
Molecular Weight:
469.41
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing