Roche Life Science Products
Breakthroughs are driven not only by inquisition and keen insight into biological processes but also by the molecular tools currently available. Progressing your research often employs a multifaceted approach utilizing genomics, proteomics, and cellular analyses. In order to enable your next discovery, we exclusively offer the complete Roche portfolio of biochemical reagents, as well as a comprehensive range of their qPCR and nucleic acid purification products. Through the spirit of collaborative innovation, our partnership with Roche includes our combined rich legacies and drive for continued product quality excellence to truly offer a seamless experience and a world-class suite of advanced molecular tools. For additional information on the power of our unique collaboration with Roche, please visit our Roche distribution agreement page.
Section Overview
Products
As pioneers and current leaders in the field of PCR, Roche nucleic acid purification, PCR and qPCR products provide you with assurance that your research is valid from the start. Additionally, our suite of reagents for DNA transfection, cell isolation and function analyses, and our protein protease and phosphatase inhibitors provide end-to-end solutions throughout your gene and protein expression workflows. We invite you to explore our combined offerings and discover how we can help you drive your next discovery.
Genomics Research
PCR, qPCR, RT-PCR, RT-qPCR, and DIG-Labeling Reagents and Protocols
An essential technique within virtually every molecular biology lab, polymerase chain reaction (PCR) is an easy and affordable method for amplifying specific fragments of DNA by several orders of magnitude. Our specialized kits have been optimized for a variety of PCR applications. Enabling room temperature PCR reaction setup with hot start PCR reagents, FastStart™ Taq DNA Polymerase is also compatible with automated setups.
Whether you need Kapa Biosystems PCR products for DNA extraction and amplification, or Kapa qPCR and qRT-PCR products utilizing SYBR® Green reagents, we have your PCR needs covered. The SYBR® Green-based qPCR reagent uses fluorescent dye that binds to double-stranded DNA molecules in order for researchers to quantify the amount of DNA that has been amplified. Probe-based qPCR reagents, such as FastStart™ Universal Probe Master (ROX), is suitable for use on all real-time PCR instruments that normalize with ROX. For RT-PCR/qRT-PCR applications, the Transcriptor One-Step RT-PCR Kit is designed for fast, sensitive, and specific end-point RT-PCR analysis of RNA. Additional nucleic acid labeling applications are readily met with the DIG-System for membrane hybridization and in situ hybridization needs.
Proteomics Analysis
Protein Stabilization Reagents and Protocols
It is essential to prevent the degradation or aggregation of the protein of interest as those factors will likely negatively impact the protein’s overall structure and functionality. This can be achieved through the addition of protease and phosphatase inhibitors to buffers and solutions. Widely cited, cOmplete™ and PhosSTOP™ protease and phosphatase inhibitors from Roche are easy-to-use, flexible tablet-based formulations which inhibit a wide range of proteases and phosphatases respectively. With the cOmplete™ Protease Inhibitor Cocktail, each of the water-soluble tablets contains a broad blend of metalloproteases, serine, and cysteine protease inhibitors to ensure the protection of proteins isolated from almost any tissue or cell. Additionally, the PhosSTOP™ Phosphatase Inhibitor Cocktail tablet provides a quick-dissolving, water-soluble formulation that contains a proprietary blend of acid and alkaline phosphatase inhibitors, serine/threonine phosphatase inhibitors and tyrosine protein phosphatase inhibitors.
Cell Research
Cell Function, Cell Isolation, and Transfection Reagents and Protocols
Involving the introduction of DNA or RNA into eukaryotic cells, transfection is a widely used technique for the study and modulation of gene expression. Created to afford high transfection efficiency, ease-of-use and reproducibility, X-tremeGENE™ Transfection Reagents provide increased viable cells and more physiologically relevant results for both transfection and siRNA reagents to knock-down gene expression. While many techniques are used to generate data from heterogeneous cell populations, it is often more informative to separate and analyze individual cellular sub-types. Collagenase and Liberase™ Research Grade enzymes are suitable for use with a broad range of cell and tissue types. Additionally, understanding cellular function also highly informative for evaluating cellular health or suggesting whether a potential drug candidate may be cytotoxic. The Roche biochemical reagents portfolio includes optimized ELISA kits for the measurement of cellular proliferation and cell death.
Brand® GmbH and BrandTech® glassware and plasticware, lab storage solutions, bottle-top dispensers, pipettes and pipette tips, cuvettes, and cell culture and PCR consumables for laboratories.
Corning® brand glass and plastic labware, filtration membranes, cell culture supplies, and equipment for life sciences.
Cytiva™ products for drug discovery research and therapeutics bioprocess manufacturing including Whatman® lab filtration products, Amersham™ chromatography columns and resins, tissue culture reagents, and products for protein sample preparation, blotting, and analysis.
Eppendorf® brand conical and microfuge tubes, pipettes and pipette tips, dispensers, centrifuges, microtiter plates, mixers, spectrometers, and PCR equipment for laboratories.
Related Product Resources
- Tissue Dissociation Guide: Collagenase, Dispase, and Liberase Enzyme Types
Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.
- Roche PCR Reagents and PCR Protocols
Comprehensive selection guide to Roche polymerase chain reaction (PCR) reagents and their applications.
- Roche Protein Stabilization Reagents
Get maximum protection after protein isolation by using protease and phosphatase inhibitor cocktail tablets from Roche.
- Digoxigenin (DIG) Labeling Methods
Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.
- X-tremeGENE™ Transfection Reagents Comparison Guide
Explore our X-tremeGENE™ transfection reagent recommendations guide for the cell type and application best suited for your specific needs.
- Hot Start PCR
The purpose of Hot Start PCR is to inhibit the PCR reaction in order to reduce nonspecific amplification, prevent the formation of primer dimers, and increase product yields.