D2622LMSUA
Sulfur in light mineral oil, ASTM D2622
Calibration Set A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
application(s)
petroleum
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
Components
Set A
Blank, 0.0005% (5ppm), 0.001% (10ppm), 0.01% (100ppm), 0.025% (250ppm), 0.05% (500ppm), 0.075% (750ppm), 0.1% (1000ppm); 100 mL ea.
Blank, 0.0005% (5ppm), 0.001% (10ppm), 0.01% (100ppm), 0.025% (250ppm), 0.05% (500ppm), 0.075% (750ppm), 0.1% (1000ppm); 100 mL ea.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
170.6 °F - closed cup
Flash Point(C)
77 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service