TBN15
Total Base Number Standard
ASTM D2896 (15.17 mg KOH/g), pkg of 1 × 50 g
Synonym(s):
TBN-Standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
according to ASTM® D2896
shelf life
limited shelf life, expiry date on the label
packaging
pkg of 1 × 50 g
manufacturer/tradename
Paragon Scientific Ltd
application(s)
petroleum
format
mixture
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Total Acid Number Standards can be found on the Paragon Scientific webpage
Application
This CRM is intended for the calibration and validation of analytical instruments used to measure base number by potentiometric titration.
Features and Benefits
- Manufactured and verified in an ISO 17025 and ISO 17034 dual accredited laboratory, as per UKAS
- Fully traceable to recognized national standards, and to units of measurement recognized at the National Institute Standards and Technology (NIST) and other recognized national standards laboratories
- Certified in accordance with ASTM D2896 and other equivalent procedures of IP 276, ISO 3771, and BS 2000 Part 276
- Typically used in, but not limited to, the analysis of used oils and lubricants
- Available in a single-volume pack of 50 g or as triple packs of the same size
- Supplied in tamper-evident glass packaging, to assure sample integrity
- Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service